Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glyctk
Entrez Gene ID
570678
Gene Name
glycerate kinase
Gene Aliases
zgc:153346
Chromosome Location
Chr.6: 43019759 - 43051634 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC124662.1 - 2 - 3 556 96

View Full Product Details