Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myf6
Entrez Gene ID
404208
Gene Name
myogenic factor 6
Gene Aliases
herculin, mrf4
Chromosome Location
Chr.4: 21717793 - 21720943 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003982.1 NP_001003982.1 1 - 2 509 80
GenBank mRNA AY335193.1 - 1 - 2 509 80

View Full Product Details