Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fdx2
Entrez Gene ID
767726
Gene Name
ferredoxin 2
Gene Aliases
fdx1l, zgc:153554
Chromosome Location
Chr.6: 149405 - 152955 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076664.2 NP_001070132.1 1 - 2 212 67
GenBank mRNA BC124384.1 - 1 - 2 193 67
EH493611.1 - 1 - 2 259 67

View Full Product Details