Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps3a
Entrez Gene ID
337240
Gene Name
ribosomal protein S3A
Gene Aliases
fb02h01, rpS3Ae, wu:fb02h01, zgc:73195, zgc:86672
Chromosome Location
Chr.1: 24355521 - 24360249 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200059.2 NP_956353.1 4 - 5 588 68
GenBank mRNA AY648734.1 - 4 - 5 569 68
BC059543.1 - 4 - 5 567 68
BC078649.1 - 4 - 5 588 68
CK357107.1 - 4 - 5 602 68

View Full Product Details