Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nop10
Entrez Gene ID
445391
Gene Name
NOP10 ribonucleoprotein homolog (yeast)
Gene Aliases
nola3, zgc:109956
Chromosome Location
Chr.7: 1505492 - 1508918 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003868.2 NP_001003868.1 2 - 2 251 60
XM_021477469.1 XP_021333144.1 2 - 2 266 60
XM_021477470.1 XP_021333145.1 2 - 2 266 60
GenBank mRNA AY648784.1 - 2 - 2 203 60
BC095081.1 - 2 - 2 251 60

View Full Product Details