Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnasekb
Entrez Gene ID
336290
Gene Name
ribonuclease, RNase K b
Gene Aliases
fj61g06, wu:fj61g06, zgc:153424
Chromosome Location
Chr.10: 22916113 - 22918214 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045375.1 NP_001038840.1 1 - 1 42 89
GenBank mRNA BC122286.1 - 1 - 1 42 89

View Full Product Details