Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chodl
Entrez Gene ID
550240
Gene Name
chondrolectin
Gene Aliases
zgc:110088
Chromosome Location
Chr.24: 24015030 - 24028235 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017578.2 NP_001017578.1 6 - 7 877 62
GenBank mRNA BC092727.1 - 6 - 7 879 62
BC155240.1 - 6 - 7 1025 62
BC164239.1 - 6 - 7 843 62
JQ790525.1 - 6 - 7 949 62

View Full Product Details