Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zbtb24
Entrez Gene ID
553955
Gene Name
zinc finger and BTB domain containing 24
Gene Aliases
zfp450, zgc:194556
Chromosome Location
Chr.17: 59785 - 77702 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001023572.2 NP_001018866.2 2 - 3 508 110
GenBank mRNA BC163252.1 - 1 - 2 447 110

View Full Product Details