Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stmn2b
Entrez Gene ID
100003563
Gene Name
stathmin 2b
Gene Aliases
scg10, scgn10, stmn2, zgc:110132
Chromosome Location
Chr.19: 31771252 - 31790551 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024222.1 NP_001019393.1 2 - 3 154 72
XM_005159590.4 XP_005159647.1 2 - 3 367 72
GenBank mRNA BC095180.1 - 2 - 3 154 72

View Full Product Details