Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan13a
Entrez Gene ID
798836
Gene Name
tetraspanin 13a
Gene Aliases
zgc:91858
Chromosome Location
Chr.15: 34592215 - 34599773 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002334.1 NP_001002334.1 3 - 4 388 100
GenBank mRNA BC076516.1 - 3 - 4 388 100
BC165139.1 - 3 - 4 341 100

View Full Product Details