Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tle3a
Entrez Gene ID
30206
Gene Name
TLE family member 3, transcriptional corepressor a
Gene Aliases
grg2, gro2, groucho2, sb:eu295, sb:eu753
Chromosome Location
Chr.18: 231350 - 261261 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313694.1 NP_001300623.1 10 - 11 920 77
NM_131012.2 NP_571087.2 10 - 11 917 77
XM_005159039.4 XP_005159096.1 11 - 12 1305 77
XM_005159041.4 XP_005159098.1 11 - 12 1302 77
XM_017351898.2 XP_017207387.1 11 - 12 1302 77
GenBank mRNA BC162533.1 - 10 - 11 818 77
U96340.1 - 10 - 11 820 77
Y12466.1 - - 820 77

View Full Product Details