Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pvalb2
Entrez Gene ID
58028
Gene Name
parvalbumin 2
Gene Aliases
cb391, ns:zf-a341, pvalb, pvalbl, zf-a341, zgc:111925
Chromosome Location
Chr.12: 154938 - 157011 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131516.2 NP_571591.1 4 - 5 450 114
GenBank mRNA AF180888.1 - 4 - 5 331 114
BC093135.1 - 4 - 5 450 114

View Full Product Details