Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
p2rx4b
Entrez Gene ID
387301
Gene Name
purinergic receptor P2X, ligand-gated ion channel, 4b
Gene Aliases
p2rx4.2, p2xr4.2
Chromosome Location
Chr.8: 345788 - 349936 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198987.2 NP_945338.2 11 - 12 1180 154
XM_021478206.1 XP_021333881.1 11 - 12 1632 154
GenBank mRNA AY292652.1 - 11 - 12 1175 154
BC162774.1 - 11 - 12 1180 154

View Full Product Details