Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
josd2
Entrez Gene ID
393120
Gene Name
Josephin domain containing 2
Gene Aliases
zgc:55937
Chromosome Location
Chr.16: 233283 - 257019 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200151.2 NP_956445.2 4 - 5 628 92
GenBank mRNA BC044546.1 - 3 - 4 549 92
EH536544.1 - 4 - 5 658 92

View Full Product Details