Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vipb
Entrez Gene ID
100000558
Gene Name
vasoactive intestinal peptide b
Gene Aliases
im:7156614, vip2
Chromosome Location
Chr.11: 43401571 - 43414160 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114555.1 NP_001108027.1 3 - 4 268 81
XM_021479523.1 XP_021335198.1 3 - 4 335 81
GenBank mRNA EU031790.1 - 3 - 4 268 81

View Full Product Details