Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prom1a
Entrez Gene ID
322857
Gene Name
prominin 1a
Gene Aliases
fb75c01, prom1, proml1, wu:fb75c01
Chromosome Location
Chr.14: 46456088 - 46616591 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115143.2 NP_001108615.2 1 - 2 315 65
XM_005157347.3 XP_005157404.1 1 - 2 440 65
XM_005157349.3 XP_005157406.1 1 - 2 441 65
XM_005157351.3 XP_005157408.1 1 - 2 441 65
XM_009291267.3 XP_009289542.1 1 - 2 543 65
XM_009291269.3 XP_009289544.1 1 - 2 440 65
XM_021480968.1 XP_021336643.1 1 - 2 544 65
XM_021480969.1 XP_021336644.1 1 - 2 544 65
XM_021480970.1 XP_021336645.1 1 - 2 544 65
XM_021480971.1 XP_021336646.1 1 - 2 543 65
XM_021480972.1 XP_021336647.1 1 - 2 543 65
XM_021480973.1 XP_021336648.1 1 - 2 441 65
XM_021480974.1 XP_021336649.1 1 - 2 544 65
XM_021480975.1 XP_021336650.1 1 - 2 543 65
GenBank mRNA AF160970.1 - 1 - 2 440 65
BC163446.1 - 1 - 2 315 65
HQ386793.1 - 1 - 2 445 65
HQ386794.1 - 1 - 2 448 65
HQ386795.1 - 1 - 2 440 65
HQ386796.1 - 1 - 2 452 65

View Full Product Details