Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sec22bb
Entrez Gene ID
327277
Gene Name
SEC22 homolog B, vesicle trafficking protein b
Gene Aliases
sec22l1b, wu:fd20b06
Chromosome Location
Chr.8: 19504011 - 19512270 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201302.2 NP_958459.1 1 - 2 212 67
GenBank mRNA BC055515.1 - 1 - 2 168 67
EH489983.1 - 1 - 2 201 67

View Full Product Details