Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdpk1b
Entrez Gene ID
403000
Gene Name
3-phosphoinositide dependent protein kinase 1b
Gene Aliases
zgc:77318
Chromosome Location
Chr.24: 37301852 - 37326125 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205699.1 10 - 11 1122 79
GenBank mRNA BC065959.1 - 10 - 11 1122 79

View Full Product Details