Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rps2
- Entrez Gene ID
-
406565
- Gene Name
- ribosomal protein S2
- Gene Aliases
-
fc94f01, ik:tdsubc_2h2, rps2e, tdsubc_2h2, wu:fb10e01, wu:fc94f01, wu:fj09b04, xx:tdsubc_2h2, zgc:85824
- Chromosome Location
-
Chr.3: 18398842 - 18403177 on Build GRCz11
- Gene Symbol
- rpl6
- Entrez Gene ID
-
336727
- Gene Name
- ribosomal protein L6
- Gene Aliases
-
wu:fa93h05, wu:fb26a11, zgc:111805
- Chromosome Location
-
Chr.8: 2493794 - 2506356 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC153958.1 |
- |
- |
767 |
147 |
View Full Product Details