Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNT1
Entrez Gene ID
100033893
Gene Name
cyclin T1
Gene Aliases
CYCT1
Chromosome Location
Chr.6: 67467526 - 67491804 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081846.1 NP_001075315.1 2 - 3 234 144
GenBank mRNA AF137509.1 - 2 - 3 234 144
AF190905.1 - 2 - 3 234 144

View Full Product Details