Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP90AA1
Entrez Gene ID
100033837
Gene Name
heat shock protein 90 alpha family class A member 1
Gene Aliases
HSPCA
Chromosome Location
Chr.24: 44233330 - 44239087 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163955.1 NP_001157427.1 1 - 2 156 123
XM_023627692.1 XP_023483460.1 - 334 123
GenBank mRNA AB043677.1 - 1 - 2 123 123

View Full Product Details