Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSP90AA1
Entrez Gene ID
100033837
Gene Name
heat shock protein 90 alpha family class A member 1
Gene Aliases
HSPCA
Chromosome Location
Chr.24: 44233330 - 44239087 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163955.1 NP_001157427.1 8 - 9 1758 107
XM_023627692.1 XP_023483460.1 - 1936 107
GenBank mRNA AB043677.1 - 8 - 9 1725 107
JL617688.1 - 6 - 7 1210 107

View Full Product Details