Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNLIPRP2
Entrez Gene ID
100033913
Gene Name
pancreatic lipase related protein 2
Gene Aliases
PLRP2
Chromosome Location
Chr.1: 15601502 - 15621477 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014732990.2 XP_014588476.2 - 1484 91
XM_023638775.1 XP_023494543.1 - 1426 91
GenBank mRNA AF359386.1 - 10 - 11 1297 91

View Full Product Details