Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPARA
Entrez Gene ID
100049840
Gene Name
peroxisome proliferator activated receptor alpha
Gene Aliases
-
Chromosome Location
Chr.28: 43180883 - 43252049 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242553.1 NP_001229482.1 4 - 5 710 77
XM_023630977.1 XP_023486745.1 - 998 77
XM_023630978.1 XP_023486746.1 - 1122 77
XM_023630979.1 XP_023486747.1 - 991 77
XM_023630980.1 XP_023486748.1 - 1165 77
XM_023630981.1 XP_023486749.1 - 1051 77
XM_023630982.1 XP_023486750.1 - 1053 77
XM_023630983.1 XP_023486751.1 - 308 77
XM_023630984.1 XP_023486752.1 - 829 77

View Full Product Details