Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKAP17A
Entrez Gene ID
418664
Gene Name
A-kinase anchoring protein 17A
Gene Aliases
SFRS17A
Chromosome Location
Chr.1: 130340807 - 130350924 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025149894.1 XP_025005662.1 1 - 2 226 75
GenBank mRNA BX931250.1 - 2 - 3 311 75

View Full Product Details