Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
428770
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4H4ORF21
Chromosome Location
Chr.4: 57097810 - 57121879 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015276340.2 XP_015131826.1 22 - 23 4723 82
XM_015276342.2 XP_015131828.1 22 - 23 4681 82
GenBank mRNA CR406058.1 - 1 - 2 70 82
CR406252.1 - 2 - 3 164 82

View Full Product Details