Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT16
Entrez Gene ID
427856
Gene Name
Wnt family member 16
Gene Aliases
-
Chromosome Location
Chr.1: 23385216 - 23397661 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001233677.5 XP_001233678.4 3 - 4 853 63
GenBank mRNA AY753296.1 - 1 - 2 286 63

View Full Product Details