Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL14
Entrez Gene ID
427926
Gene Name
F-box and leucine rich repeat protein 14
Gene Aliases
PPA
Chromosome Location
Chr.1: 60881396 - 60885783 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015286684.2 XP_015142170.1 1 - 1 1861 92
GenBank mRNA CR391509.1 - 1 - 1 853 92

View Full Product Details