Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL19
Entrez Gene ID
427406
Gene Name
C-C motif chemokine ligand 19
Gene Aliases
-
Chromosome Location
Chr.Z: 8363698 - 8366694 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302168.1 NP_001289097.1 2 - 3 721 150
GenBank mRNA BX929857.2 - 2 - 3 731 150

View Full Product Details