Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCTN5
Entrez Gene ID
416574
Gene Name
dynactin subunit 5
Gene Aliases
-
Chromosome Location
Chr.14: 7289000 - 7295773 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198639.1 NP_001185568.1 1 - 2 69 73
NM_001198640.1 NP_001185569.1 1 - 2 69 73
GenBank mRNA BX934787.1 - 1 - 2 54 73
BX935500.2 - 1 - 2 55 73
CN235561.1 - 1 - 2 44 73
CR353698.1 - 1 - 2 51 73

View Full Product Details