Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM21C
Entrez Gene ID
423772
Gene Name
family with sequence similarity 21 member C
Gene Aliases
FAM21A, WASHC2C
Chromosome Location
Chr.6: 18616583 - 18647713 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012593.2 NP_001012611.2 17 - 18 1632 116
XM_015288386.2 XP_015143872.1 17 - 18 1702 116
XM_015288389.2 XP_015143875.1 17 - 18 1702 116
XM_015288390.2 XP_015143876.1 17 - 18 1702 116
XM_025151585.1 XP_025007353.1 17 - 18 1702 116
XM_025151586.1 XP_025007354.1 17 - 18 1702 116
XM_025151587.1 XP_025007355.1 17 - 18 1702 116
XM_025151588.1 XP_025007356.1 16 - 17 1645 116
XM_025151589.1 XP_025007357.1 16 - 17 1528 116
GenBank mRNA AJ851808.1 - 17 - 18 1631 116

View Full Product Details