Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPGD
Entrez Gene ID
422567
Gene Name
hydroxyprostaglandin dehydrogenase 15-(NAD)
Gene Aliases
-
Chromosome Location
Chr.4: 43759685 - 43792292 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277784.1 NP_001264713.1 2 - 3 339 71
XM_025149634.1 XP_025005402.1 3 - 4 1522 71
GenBank mRNA BU258980.1 - 2 - 3 347 71
BX929847.1 - 2 - 3 347 71
BX934937.1 - 2 - 3 287 71
CR353827.1 - 2 - 3 71

View Full Product Details