Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPGD
Entrez Gene ID
422567
Gene Name
hydroxyprostaglandin dehydrogenase 15-(NAD)
Gene Aliases
-
Chromosome Location
Chr.4: 43759685 - 43792292 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277784.1 NP_001264713.1 3 - 4 453 99
XM_025149634.1 XP_025005402.1 4 - 5 1636 99
GenBank mRNA BU258980.1 - 3 - 4 461 99
BX929847.1 - 3 - 4 461 99
BX934937.1 - 3 - 4 401 99
CR353827.1 - 3 - 4 375 99

View Full Product Details