Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPGD
Entrez Gene ID
422567
Gene Name
hydroxyprostaglandin dehydrogenase 15-(NAD)
Gene Aliases
-
Chromosome Location
Chr.4: 43759685 - 43792292 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277784.1 NP_001264713.1 5 - 6 621 115
XM_015276239.2 XP_015131725.1 3 - 4 294 115
XM_025149634.1 XP_025005402.1 6 - 7 1804 115
GenBank mRNA BX929847.1 - 5 - 6 629 115
BX934937.1 - 5 - 6 569 115
CR353827.1 - 5 - 6 543 115

View Full Product Details