Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC107049082
Entrez Gene ID
107049082
Gene Name
sodium/potassium-transporting ATPase subunit beta-1-interacting protein 3-like
Gene Aliases
-
Chromosome Location
Chr.2: 113438537 - 113770112 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015282718.2 XP_015138204.2 4 - 5 663 92
XM_025147985.1 XP_025003753.1 4 - 5 706 92
GenBank mRNA BX931733.2 - 4 - 5 714 92

View Full Product Details