Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLRT3
Entrez Gene ID
428552
Gene Name
fibronectin leucine rich transmembrane protein 3
Gene Aliases
-
Chromosome Location
Chr.3: 4681080 - 4694794 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX934700.2 - 2 - 3 234 86

View Full Product Details