Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASTKD5
Entrez Gene ID
422937
Gene Name
FAST kinase domains 5
Gene Aliases
-
Chromosome Location
Chr.4: 88667561 - 88674248 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_420875.6 XP_420875.5 2 - 2 1061 97
GenBank mRNA BX933346.2 - 1 - 1 1048 97

View Full Product Details