Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLPH3
Entrez Gene ID
427422
Gene Name
golgi phosphoprotein 3
Gene Aliases
-
Chromosome Location
Chr.Z: 9664867 - 9697214 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_424995.6 XP_424995.3 1 - 2 493 60
GenBank mRNA CR406158.1 - 1 - 2 72 60

View Full Product Details