Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXT2
Entrez Gene ID
422346
Gene Name
nuclear transport factor 2 like export factor 2
Gene Aliases
-
Chromosome Location
Chr.4: 13807631 - 13813922 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006436.1 NP_001006436.1 2 - 3 127 119
GenBank mRNA AJ719764.1 - 2 - 3 127 119

View Full Product Details