Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SP3
Entrez Gene ID
395302
Gene Name
Sp3 transcription factor
Gene Aliases
-
Chromosome Location
Chr.7: 17080571 - 17112068 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204603.1 NP_989934.1 4 - 5 1809 77
XM_015289377.2 XP_015144863.1 5 - 6 1922 77
XM_025152088.1 XP_025007856.1 2 - 3 1627 77
GenBank mRNA AJ317961.1 - 4 - 5 1809 77
AJ719471.1 - 2 - 3 1457 77

View Full Product Details