Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPN1MSW
Entrez Gene ID
396486
Gene Name
opsin, green sensitive (rhodopsin-like)
Gene Aliases
CHK-1, OPNG, PRA1, RHO
Chromosome Location
Chr.26: 4556844 - 4562454 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205490.1 NP_990821.1 - 648 62
GenBank mRNA M92038.1 - - 648 62

View Full Product Details