Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
396499
Gene Name
crystallin beta A1
Gene Aliases
-
Chromosome Location
Chr.19: 6105041 - 6114864 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205502.2 NP_990833.2 3 - 4 223 60
XM_015295686.2 XP_015151172.1 6 - 7 760 60
XM_015295687.2 XP_015151173.1 5 - 6 1396 60
XM_015295688.2 XP_015151174.1 6 - 7 647 60
XM_015295690.2 XP_015151176.1 4 - 5 1275 60
XM_025141928.1 XP_024997696.1 4 - 5 1785 60
XM_025141929.1 XP_024997697.1 5 - 6 1283 60
XM_025141930.1 XP_024997698.1 5 - 6 553 60
GenBank mRNA DR424139.1 - 3 - 4 210 60
M15658.1 - 3 - 4 223 60

View Full Product Details