Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRYBA1
Entrez Gene ID
396499
Gene Name
crystallin beta A1
Gene Aliases
-
Chromosome Location
Chr.19: 6105041 - 6114864 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205502.2 NP_990833.2 5 - 5 435 118
XM_015295686.2 XP_015151172.1 8 - 8 972 118
XM_015295687.2 XP_015151173.1 7 - 7 1608 118
XM_015295688.2 XP_015151174.1 8 - 8 859 118
XM_015295690.2 XP_015151176.1 6 - 6 1487 118
XM_025141928.1 XP_024997696.1 6 - 6 1997 118
XM_025141929.1 XP_024997697.1 7 - 7 1495 118
XM_025141930.1 XP_024997698.1 7 - 7 765 118
GenBank mRNA DR424139.1 - 5 - 5 422 118
M15658.1 - 5 - 5 435 118

View Full Product Details