The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012948.1 | NP_001012966.1 | 1 - 2 | 2913 | 89 |
GenBank mRNA | AJ720863.1 | - | 1 - 2 | 2913 | 89 |