Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATA2
Entrez Gene ID
416018
Gene Name
GATA binding protein 2
Gene Aliases
GATA-2, NF-E1b
Chromosome Location
Chr.12: 9596109 - 9612566 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003797.1 NP_001003797.1 3 - 4 1239 58
XM_015293079.2 XP_015148565.1 3 - 4 1312 58
GenBank mRNA X56930.1 - - 1239 58

View Full Product Details