Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CECR1
Entrez Gene ID
418160
Gene Name
cat eye syndrome chromosome region, candidate 1
Gene Aliases
ADA2
Chromosome Location
Chr.1: 61653446 - 61672321 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030767.2 NP_001025938.1 6 - 7 1082 134
XM_015285830.2 XP_015141316.1 6 - 7 1639 134
XM_015285839.2 XP_015141325.1 6 - 7 1096 134
GenBank mRNA AJ720326.1 - 6 - 7 1083 134
AY902779.1 - 6 - 7 1085 134

View Full Product Details