Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF3H
Entrez Gene ID
420288
Gene Name
eukaryotic translation initiation factor 3 subunit H
Gene Aliases
EIF3S3
Chromosome Location
Chr.2: 135418684 - 135499641 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030951.1 NP_001026122.1 2 - 3 306 142
GenBank mRNA AJ719788.1 - 2 - 3 306 142

View Full Product Details