Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R2
Entrez Gene ID
424894
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 2
Gene Aliases
-
Chromosome Location
Chr.9: 12836900 - 12849719 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031313.1 NP_001026484.1 5 - 6 654 108
GenBank mRNA AJ851823.1 - 5 - 6 654 108

View Full Product Details