Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDSUB1
Entrez Gene ID
424331
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 36520588 - 36550929 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031263.2 NP_001026434.2 10 - 11 1617 97
XM_015290154.1 XP_015145640.1 9 - 10 1689 97
XM_015290155.1 XP_015145641.1 9 - 10 1689 97
XM_015290156.1 XP_015145642.1 9 - 10 2320 97
XM_015290160.2 XP_015145646.1 11 - 12 1776 97
XM_015290161.2 XP_015145647.1 11 - 12 1747 97
XM_015290162.2 XP_015145648.1 11 - 12 1921 97
XM_015290163.2 XP_015145649.1 10 - 11 1659 97
XM_015290164.2 XP_015145650.1 10 - 11 1634 97
XM_015290165.2 XP_015145651.1 10 - 11 1394 97
XM_015290166.2 XP_015145652.1 11 - 12 1543 97
XM_015290167.2 XP_015145653.1 10 - 11 1186 97
XM_025152249.1 XP_025008017.1 9 - 10 1656 97
XM_025152250.1 XP_025008018.1 11 - 12 1885 97
XM_025152252.1 XP_025008020.1 11 - 12 1960 97
XM_025152253.1 XP_025008021.1 11 - 12 1889 97
XM_025152254.1 XP_025008022.1 11 - 12 1751 97
XM_025152255.1 XP_025008023.1 11 - 12 1801 97
XM_025152256.1 XP_025008024.1 11 - 12 1511 97
XM_025152257.1 XP_025008025.1 11 - 12 1805 97
XM_025152258.1 XP_025008026.1 10 - 11 1193 97
GenBank mRNA AJ719461.1 - 10 - 11 1619 97
AJ726212.1 - 1 - 2 105 97
BU364244.1 - 6 - 7 607 97

View Full Product Details