Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CELA2A
Entrez Gene ID
419486
Gene Name
chymotrypsin like elastase family member 2A
Gene Aliases
ELA2A
Chromosome Location
Chr.21: 5543786 - 5550474 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032390.2 NP_001027562.2 6 - 7 664 87
XM_025142533.1 XP_024998301.1 2 - 3 496 87
GenBank mRNA BU487365.1 - 3 - 4 355 87
BX933628.2 - 5 - 6 542 87
BX933648.1 - 6 - 7 646 87
BX933649.2 - 6 - 7 646 87
BX933653.2 - 6 - 7 661 87
BX933662.1 - 6 - 7 629 87
BX933680.2 - 6 - 7 638 87
BX933686.1 - 6 - 7 664 87
BX935221.1 - 6 - 7 634 87
BX935323.2 - 6 - 7 646 87
BX935324.2 - 6 - 7 638 87
BX935330.2 - 6 - 7 626 87
CR338993.2 - 6 - 7 646 87

View Full Product Details